Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF30 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524104

Catalog No. PIPA524104


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF30
Polyclonal
PBS with 0.09% sodium azide
P17039
KOX28, zinc finger protein 30 (KOX 28), zinc finger protein KOX28
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90075
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ZNF30
Liquid
ZNF30
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ZNF30
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit