Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF313 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525600

Catalog No. PIPA525600

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ZNF313 is a novel human zinc finger protein gene that contains both ring finger and C(2)H(2) domain. It may play a role in spermatogenesis.
Specifications

Specifications

ZNF313
Polyclonal
PBS with 0.09% sodium azide
Q9Y508
PSORS12, ZNF313, RING finger protein 114, zinc finger protein 228, zinc finger protein 313
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55905
Immunohistochemistry, Western Blot
Unconjugated
RNF114
Liquid
RNF114
KLH conjugated synthetic peptide between 167-193 amino acids from the C-terminal region of human ZNF313
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit