Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF322A Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Goat, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180365

Catalog No. NBP180365


Add to cart

Description

Description

ZNF322A Polyclonal antibody specifically detects ZNF322A in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Goat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

ZNF322A
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
Affinity Purified
ZNF322
Synthetic peptide directed towards the C terminal of human ZNF322A. Peptide sequence YTVCDKSFHQSSALLQHQTVHIGEKPFVCNVSEKGLELSPPHASEASQMS.
Immunogen affinity purified
RUO
Primary
79692
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_078915
FLJ23393, HCG12, HLA complex group 12, zinc finger protein 322, zinc finger protein 322A, zinc finger protein 388, zinc finger protein 489, ZNF322A, ZNF388, ZNF489
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Equine: 100%; Bovine: 84%.
Bovine, Canine, Equine, Goat, Human, Mouse, Porcine, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only