Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF323 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527894

Catalog No. PIPA527894


Add to cart

Description

Description

PA5-27895 targets RISC in IF and WB applications and shows reactivity with Human samples. The PA5-27895 immunogen is recombit fragment corresponding to a region within amino acids 165 and 452 of Human RISC.

ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 [PubMed 12147252]).
Specifications

Specifications

ZNF323
Polyclonal
Unconjugated
ZSCAN31
Liquid
ZSCAN31
Recombit fragment corresponding to a region within amino acids 66 and 379 of Human ZNF323
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q96LW9
ZNF20-Lp, ZNF310P, dJ874C20.2, zinc finger protein 310 pseudogene
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64288
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit