Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF324B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524105

Catalog No. PIPA524105


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF324B
Polyclonal
PBS with 0.09% sodium azide
Q6AW86
ZNF324B
KLH conjugated synthetic peptide between 507-535 amino acids from the C-terminal region of human ZNF324B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ZNF324B
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
388569
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit