Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 354A (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ZNF354A.

Manufacturer:  Abnova Corporation H00006940A01

Catalog No. 89-004-334


Add to cart

Description

Description

Sequence: GLILKRSNRNVPWDLKLEKPYIYEGRLEKKQDKKGSFQIVSATHKKIPTIERSHKNTELSQNFSPKSVLIRQQILPREKTPPKCEIQGNSLKQNSQLLNQPKITADKRYK
Specifications

Specifications

zinc finger protein 354A
Unconjugated
50% glycerol
NM_005649
EZNF|HKL1|KID-1|KID1|TCF17
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant ZNF354A.
ZNF354A
Serum
ZNF354A
ZNF354A (NP_005640, 106 a.a. ~ 215 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6940
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit