Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF366 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524242

Catalog No. PIPA524242

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF366
Polyclonal
PBS with 0.09% sodium azide
Q8N895
DCSCRIPT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
167465
Western Blot
Unconjugated
ZNF366
Liquid
ZNF366
KLH conjugated synthetic peptide between 650-678 amino acids from the C-terminal region of human ZNF366
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit