Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF396 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528388

Catalog No. PIPA528388


Add to cart

Description

Description

PA5-28388 targets ZNF396 in WB applications and shows reactivity with Human samples. The PA5-28388 immunogen is recombit fragment corresponding to a region within amino acids 1 and 259 of Human ZNF396. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
Specifications

Specifications

ZNF396
Polyclonal
Unconjugated
ZNF396
Liquid
ZNF396
Recombit fragment corresponding to a region within amino acids 1 and 259 of Human ZNF396
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.94 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q96N95
ZSCAN14, zinc finger and SCAN domain-containing protein 14
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
252884
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit