Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 43, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF43 protein.

Manufacturer:  Abnova Corporation H00007594B01

Catalog No. 89-006-481


Add to cart

Description

Description

This gene belongs to the C2H2-type zinc finger gene family. The zinc finger proteins are involved in gene regulation and development, and are quite conserved throughout evolution. Like this gene product, a third of the zinc finger proteins containing C2H2 fingers also contain the KRAB domain, which has been found to be involved in protein-protein interactions. [provided by RefSeq

Sequence: MDVAIEFCLEEWQCLDIAQQNLYRNVMLENYRNLVFLGIAVSKPDLITCLEQEKEPWEPMRRHEMVAKPPVMCSHFTQDFWPEQHIKDPFQKATLRRYKNCEHKNVHLKKDHKSVDECKVHRGGYNGFNQCLPATQSKIFLFDKCVKAFHKFSNSNRHKISHTEKKLFKCKECGKSFCMLPHLAQHKIIHTRVNFCKCEKCGKAFNCPSIITKHKRINTGEKPYTCEECGKVFNWSSRLTTHKKNYTRYKLYKCEECGKAFNKSSILTTHKIIRTGEKFYKCKECAKAFNQSSNLTEHKKIHPGEKPYKCEECGKAFNWPSTLTKHKRIHTGEKPYTCEECGKAFNQFSNLTTHKRIHTAEKFYKCTECGEAFSRSSNLTKHKKIHTEKKPYKCEECGKAFKWSSKLTEHKLTHTGEKPYKCEECGKAFNWPSTLTKHNRIHTGEKPYKCEVCGKAFNQFSNLTTHKRIHTAEKPYKCEECGKAFSRSSNLTKHKKIHIEKKPYKCEECGKAFKWSSKLTEHKITHTGEKPYKCEECGKAFNHFSILTKHKRIHTGEKPYKCEECGKAFTQSSNLTTHKKIHTGEKFYKCEECGKAFTQSSNLTTHKKIHTGGKPYKCEECGKAFNQFSTLTKHKIIHTEEKPYKCEECGKAFKWSSTLTKHKIIHTGEKPYKCEECGKAFKLSSTLSTHKIIHTGEKPYKCEKCGKAFNRSSNLIEHKKIHTGEQPYKCEECGKAFNYSSHLNTHKRIHTKEQPYKCKECGKAFNQYSNLTTHNKIHTGEKLYKPEDVTVILTTPQTFSNIK
Specifications

Specifications

zinc finger protein 43
Unconjugated
No additive
BC006528
DKFZp686L1854|HTF6|KOX27|ZNF39L1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7594
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF43 protein.
ZNF43
Serum
ZNF43
ZNF43 (AAH06528, 1 a.a. ~ 803 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit