Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger and SCAN domain containing 16, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF435 protein.

Manufacturer:  Abnova Corporation H00080345B01

Catalog No. 89-008-249


Add to cart

Description

Description

Sequence: MTTALEPEDQKGLLIIKAEDHYWGQDSSSQKCSPHRRELYRQHFRKLCYQDAPGPREALTQLWELCRQWLRPECHTKEQILDLLVLEQFLSILPKDLQAWVRAHHPETGEEAVTVLEDLERELDEPGKQVPGNSERRDILMDKLAPLGRPYESLTVQLHPKKTQLEQEAGKPQRNGDKTRTKNEELFQKEDMPKDKEFLGEINDRLNKDTPQHPKSKDIIENEGRSEWQQRERRRYKCDECGKSFSHSSDLSKHRRTHTGEKPYKCDECGKAFIQRSHLIGHHRVHTGVKPYKCKECGKDFSGRTGLIQHQRIHTGEKPYECDECGRPFRVSSALIRHQRIHTANKLY
Specifications

Specifications

zinc finger and SCAN domain containing 16
Unconjugated
No additive
NM_025231.1
FLJ22191|ZNF392|ZNF435|dJ265C24.3
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF435 protein.
ZSCAN16
Serum
ZSCAN16
ZNF435 (NP_079507.1, 1 a.a. ~ 348 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
80345
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit