Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 473, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF473 protein.

Manufacturer:  Abnova Corporation H00025888B01

Catalog No. 89-007-352


Add to cart

Description

Description

Sequence: MAEEFVTLKDVGMDFTLGDWEQLGLEQGDTFWDTALDNCQDLFLLDPPRPNLTSHPDGSEDLEPLAGGSPEATSPDVTETKNSPLMEDFFEEGFSQEIIEMLSKDGFWNSNFGEACIEDTWLDSLLGDPESLLRSDIATNGESPTECKSHELKRGLSPVSTVSTGEDSMVHNVSEKTLTPAKSKEYRGEFFSYSDHSQQDSVQEGEKPYQCSECGKSFSGSYRLTQHWITHTREKPTVHQECEQGFDRNASLSVYPKTHTGYKFYVCNEYGTTFSQSTYLWHQKTHTGEKPCKSQDSDHPPSHDTQPGEHQKTHTDSKSYNCNECGKAFTRIFHLTRHQKIHTRKRYECSKCQATFNLRKHLIQHQKTHAAKTTSECQECGKIFRHSSLLIEHQALHAGEEPYKCNERGKSFRHNSTLKIHQRVHSGEKPYKCSECGKAFHRHTHLNEHRRIHTGYRPHKCQECVRSFSRPSHLMRHQAIHTAEKPYSCAECKETFSDNNRLVQHQKMHTVKTPYECQECGERFICGSTLKCHESVHAREKQGFFVSGKILDQNPEQKEKCFKCNKCEKTFSCSKYLTQHERIHTRGVKPFECDQCGKAFGQSTRLIHHQRIHSRVRLYKWGEQGKAISSASLIKLQSFHTKEHPFKCNECGKTFSHSAHLSKHQLIHAGENPFKCSKCDRVFTQRNYLVQHERTHARKKPLVCNECGKTFRQSSCLSKHQRIHSGEKPYVCDYCGKAFGLSAELVRHQRIHTGEKPYVCQECGKAFTQSSCLSIHRRVHTGEKPYRCGECGKAFAQKANLTQHQRIHTGEKPYSCNVCGKAFVLSAHLNQHLRVHTQETLYQCQRCQKAFRCHSSLSRHQRVHNKQQYCL
Specifications

Specifications

zinc finger protein 473
Unconjugated
No additive
NM_001006656.1
HZFP100|ZN473
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
25888
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF473 protein.
ZNF473
Serum
ZNF473
ZNF473 (NP_001006657.1, 1 a.a. ~ 871 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit