Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF500 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124397

Catalog No. PIPA124397


Add to cart

Description

Description

PA1-24397 detects ZNF500 in Human samples. PA1-24397 has been successfully used in Western Blot procedures. PA1-24397 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human ZNF500. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

ZNF500 is a new candidate transcription factor. It is derived from an annotated genomic sequence (NT_037887) using a gene prediction method.
Specifications

Specifications

ZNF500
Polyclonal
Unconjugated
ZNF500
Liquid
ZNF500
Synthetic peptide region of human ZNF500.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O60304
ZKSCAN18
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26048
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit