Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZNF521, FITC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191271F

Catalog No. NBP191271F


Add to cart

Description

Description

ZNF521 Polyclonal antibody specifically detects ZNF521 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

ZNF521
FITC
PBS with 0.05% Sodium Azide
Affinity Purified
ZNF521
A synthetic peptide made to an internal portion of the human ZNF521 protein (between residues 450-550) [UniProt Q96K83]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative)
Q96K83
EHZF, Evi3, LIP3, ZFP521, zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12
Rabbit
IgG
0.1 ml
Immunology, Neuroscience
Primary
25925
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only