Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 558, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF558 protein.

Manufacturer:  Abnova Corporation H00148156B01

Catalog No. 89-008-596


Add to cart

Description

Description

Sequence: MAAVILPSTAAPSSLFPASQQKGHTQGGELVNELLTSWLRGLVTFEDVAVEFTQEEWALLDPAQRTLYRDVMLENCRNLASLGCRVNKPSLISQLEQDKKVVTEERGILPSTCPDLETLLKAKWLTPKKNVFRKEQSKGVKTERSHRGVKLNECNQCFKVFSTKSNLTQHKRIHTGEKPYDCSQCGKSFSSRSYLTIHKRIHNGEKPYECNHCGKAFSDPSSLRLHLRIHTGEKPYECNQCFHVFRTSCNLKSHKRIHTGENHHECNQCGKAFSTRSSLTGHNSIHTGEKPYECHDCGKTFRKSSYLTQHVRTHTGEKPYECNECGKSFSSSFSLTVHKRIHTGEKPYECSDCGKAFNNLSAVKKHLRTHTGEKPYECNHCGKSFTSNSYLSVHKRIHNRWI
Specifications

Specifications

zinc finger protein 558
Unconjugated
No additive
NM_144693.1
FLJ30932
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
148156
Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF558 protein.
ZNF558
Serum
ZNF558
ZNF558 (NP_653294.1, 1 a.a. ~ 402 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit