Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 584, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF584 protein.

Manufacturer:  Abnova Corporation H00201514B01

Catalog No. 89-008-672


Add to cart

Description

Description

Sequence: MAGEAEAQLDPSLQGLVMFEDVTVYFSREEWGLLNVTQKGLYRDVMLENFALVSSLGLAPSRSPVFTQLEDDEQSWVPSWVDVTPVSRAEARRGFGLDGLCRVEDERAHPEHLKSYRVIQHQDTHSEGKPRRHTEHGAAFPPGSSCGQQQEVHVAEKLFKCSDCGKVFLKAFALLDHLITHSEERPFRCPTGRSAFKKSAHINPRKIHTGETAHVCNECGKAFSYPSKLRKHQKVHTGIKPFKCSDCGKTFNRKDALVLHQRIHTGERPYECSKCGKTFSVLSTLIRHRKVHIGERPYECTECGKFFKYNNSFILHQRVHTGERPFECKQCGKGYVTRSGLYQHWKVHTGERPYECSLCGKTFTTRSYRNRHQQFHTEERSYECTECGKAFKHSSTLLQHKKVHTPERRQEDRAHGKVVSC
Specifications

Specifications

zinc finger protein 584
Unconjugated
No additive
NM_173548.1
FLJ39899
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
201514
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF584 protein.
ZNF584
Serum
ZNF584
ZNF584 (NP_775819.1, 1 a.a. ~ 421 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit