Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger protein 606, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF606 protein.
Manufacturer: Abnova Corporation H00080095B01
Description
Sequence: MAAINPWASWGALTDQSWGMTAVDPWASWALCPQYPAWHVEGSLEEGRRATGLPAAQVQEPVTFKDVAVDFTQEEWGQLDLVQRTLYRDVMLETYGHLLSVGNQIAKPEVISLLEQGEEPWSVEQACPQRTCPEWVRNLESKALIPAQSIFEEEQSHGMKLERYIWDDPWFSRLEVLGCKDQLEMYHMNQSTAMRQMVFMQKQVLSQRSSEFCGLGAEFSQNLNFVPSQRVSQIEHFYKPDTHAQSWRCDSAIMYADKVTCENNDYDKTVYQSIQPIYPARIQTGDNLFKCTDAVKSFNHIIHFGDHKGIHTGEKLYEYKECHQIFNQSPSFNEHPRLHVGENQYNYKEYENIFYFSSFMEHQKIGTVEKAYKYNEWEKVFGYDSFLTQHTSTYTAEKPYDYNECGTSFIWSSYLIQHKKTHTGEKPYECDKCGKVFRNRSALTKHERTHTGIKPYECNKCGKAFSWNSHLIVHKRIHTGEKPYVCNECGKSFNWNSHLIGHQRTHTGEKPFECTECGKSFSWSSHLIAHMRMHTGEKPFKCDECEKAFRDYSALSKHERTHSGAKPYKCTECGKSFSWSSHLIAHQRTHTGEKPYNCQECGKAFRERSALTKHEIIHSGIKPYECNKCGKSCSQMAHLVRHQRTHTGEKPYECNKCGKSFSQSCHLVAHRRIHTGEKPYKCNQCERSFNCSSHLIAHRRTHTGEKPYRCNECGKAFNESSSLIVHLRNHTGEKPYKCNHCEKAFCKNSSLIIHQRMHSGEKRFICSECGKAFSGHSALLQHQRNHSEEKLNSpecifications
zinc finger protein 606 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZNF606 protein. | |
ZNF606 | |
FLJ14260/KIAA1852/ZNF328 | |
Mouse | |
50 μL | |
Yes | |
80095 | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_025027.3 | |
ZNF606 | |
ZNF606 (NP_079303.2, 1 a.a. ∼ 792 a.a) full-length human protein. | |
RUO | |
Primary | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title