Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZNF703, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232139

Catalog No. NBP232139


Add to cart

Description

Description

ZNF703 Polyclonal antibody specifically detects ZNF703 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

ZNF703
1.0 mg/mL
Immunoprecipitation 2 - 10 mcg/mg lysate
NP_079345.1
FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ZNF703
The immunogen this antibody was made to, maps to a region between residue 225 to 275 of human Zinc Finger Protein 703 using the numbering given in entry NP_079345.1 (GeneID 80139).
Immunogen affinity purified
RUO
Primary
80139
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only