Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger and SCAN domain containing 12, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ZNF96 protein.

Manufacturer:  Abnova Corporation H00009753B01

Catalog No. 89-006-838


Add to cart

Description

Description

Sequence: MASTWAIQAHMDQDEPLEVKIEEEKYTTRQDWDLRKNNTHSREVFRQYFRQFCYQETSGPREALSRLRELCHQWLRPETHTKEQILELLVLEQFLTILPEELQAWVQEQHPESGEEVVTVLEDLERELDEPGEQVSVHTGEQEMFLQETVRLRKEGEPSMSLQSMKAQPKYESPELESQQEQVLDVETGNEYGNLKQEVSEEMEPHGKTSSKFENDMSKSARCGETREPEEITEEPSACSREDKQPTCDENGVSLTENSDHTEHQRICPGEESYGCDDCGKAFSQHSHLIEHQRIHTGDRPYKCEECGKAFRGRTVLIRHKIIHTGEKPYKCNECGKAFGRWSALNQHQRLHTGEKHYHCNDCGKAFSQKAGLFHHIKIHTRDKPYQCTQCNKSFSRRSILTQHQGVHTGAKPYECNECGKAFVYNSSLVSHQEIHHKEKCYQCKECGKSFSQSGLIQHQRIHTGEKPYKCDVCEKAFIQRTSLTEHQRIHTGERPYKCDKCGKAFTQRSVLTEHQRIHTGERPYKCDECGNAFRGITSLIQHQRIHTGEKPYQCDECGKAFRQRKKTSYKEILLKNHSEPQAGVNLLLSSLIPEWQSCFRRDL
Specifications

Specifications

zinc finger and SCAN domain containing 12
Unconjugated
No additive
NM_001039643.1
KIAA0426|ZFP96|ZNF29K1|ZNF305|ZNF96|dJ29K1.2
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ZNF96 protein.
ZSCAN12
Serum
ZSCAN12
ZNF96 (NP_001034732.1, 1 a.a. ~ 604 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9753
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit