Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNHIT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527598

Catalog No. PIPA527598


Add to cart

Description

Description

PA5-27598 targets ZNHIT1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-27598 immunogen is recombit fragment corresponding to a region within amino acids 1 and 154 of Human ZNHIT1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Seems to play a role in p53-mediated apoptosis induction.
Specifications

Specifications

ZNHIT1
Polyclonal
Unconjugated
ZNHIT1
Liquid
ZNHIT1
Recombit fragment corresponding to a region within amino acids 1 and 154 of Human ZNHIT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O43257
CG1I, ZNFN4A1, H_DJ0747G18.14, cyclin-G1-binding protein 1, p18 Hamlet, p18Hamlet, putative cyclin G1 interacting protein, zinc finger HIT domain-containing protein 1, zinc finger protein subfamily 4A member 1, zinc finger protein, subfamily 4A (HIT domain containing), member
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10467
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit