Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRANB1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526829

Catalog No. PIPA526829


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

ZRANB1 is a positive regulator of the Wnt signaling pathway that specifically cleaves  ft.Lys-63 ft.-linked ubiquitin chains. It acts by deubiquitinating APC protein, a negative regulator of Wnt-mediated transcription and may also modulate TNF-alpha signaling.
Specifications

Specifications

ZRANB1
Polyclonal
Unconjugated
ZRANB1
Liquid
ZRANB1
KLH conjugated synthetic peptide between 665-692 amino acids from the C-terminal region of human ZRANB1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9UGI0, Q7M760
TRABID, TRAF-binding protein domain, hTrabid, ubiquitin thioesterase ZRANB1, zinc finger Ran-binding domain-containing protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
360216, 54764
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit