Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ZRANB3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152644

Catalog No. NBP152644


Add to cart

Description

Description

ZRANB3 Polyclonal antibody specifically detects ZRANB3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ZRANB3
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg
Affinity Purified
ZRANB3
The immunogen for this product maps to a region between residue 1029 and 1079 of human Zinc Finger, RAN-binding Domain Containing 3 using the numbering given in entry NP_115519.2 (GeneID 84083).
Immunogen affinity purified
RUO
Primary
84083
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
MGC105033, MGC75012,4933425L19Rik, zinc finger Ran-binding domain-containing protein 3, zinc finger, RAN-binding domain containing 3
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only