Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zyxin Mouse anti-Chicken, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610521

Catalog No. BDB610521


Add to cart

Description

Description

Zyxin is an 82kDa zinc-binding metalloprotein found in focal adhesion plaques. It contains a proline-rich N-terminal domain and a C-terminal domain containing three LIM domains. LIM domains are characterized as zinc-binding motifs involved in gene expression protein-protein interactions. Zyxin interacts with several focal adhesion proteins including α-actinin, Vav, and another LIM domain-containing protein, cCRP. Zyxin associates selectively with an SH3 domain of p95 Vav through its proline-rich N-terminal domain. The focal adhesion protein VASP has also been shown to interact with the 83kDa human homologue of zyxin, p83. Other LIM-containing proteins function as transcription factors or as growth and differentiation factors in embryogenesis. Zyxin and its homologues appear to be involved in the transduction of extracellular signals originating at focal adhesion plaques.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Zyxin
21
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Chicken
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit