Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOJO™ Antibacterial Plum Foam Handwash

Antibacterial foam hand soap with plum fragrance.
Supplier: GOJO™ 131203
Description
- Enriched with a natural extract and skin conditioner
- USDA Certified Biobased Product
- SMART-FLEX™ bottle design uses 30% less material than HDPE
- Patent-pending controlled collapse technology maintains bottle shape longer while emptying
- Removable pump for easier recycling
- SANITARY SEALED™ refill helps prevent contamination

Specifications
Hand Soap | |
Foam | |
3/Pk. | |
700 mL |
Plum | |
GOJO™ LTX-7™ Dispenser | |
700 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction