Learn More
DWK Life Sciences™ Wheaton™ AntiBIND™ Deep Well Microplates
Used in chromatography applications, long term sample storage, and bioanalytical assays. Wheaton AntiBIND 96 Deep Well Microplates, Conical, are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding and ensures ultimate recovery of valuable proteins.
Manufacturer:  DWK Life Sciences™ W266032
 View more versions of this product
Catalog No. 02-912-404
Add to cart
Description
Description
  • Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
  • Meets the SBS and ANSI dimensions used for automation
  • Conical shaped bottoms for maximum sample recovery and efficient mixing
  • Manufactured from polypropylene resin
Specifications
Specifications
Conical
High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS)
20
2mL
Natural
96
2mL
Deep-Well Microplate