Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ ANTIBind High Purity Sealing Mats

High purity natural-colored silcone sealing mats with PTFE spray coating.

$39.00

Specifications

Material Silicone with PTFE Coating
No. per Pack 5
For Use With ANTIBind plates
Color Natural
View More Specs

Products 2
Catalog Number Mfr. No. No. of Wells Price Quantity & Availability  
Catalog Number Mfr. No. No. of Wells Price Quantity & Availability  
02-911-706 View Documents DWK Life Sciences
W266044
96 Case of 5 for $39.00
Add to cart
 
02-911-707 View Documents DWK Life Sciences
W266046
384 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • For use with ANTIBind 384- or 96-well plates
  • Packaged in 5 mats/pack
Specifications

Specifications

Silicone with PTFE Coating
5
ANTIBind plates
Natural
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit