Learn More
DWK Life Sciences Wheaton™ ANTIBind High Purity Sealing Mats
High purity natural-colored silcone sealing mats with PTFE spray coating.
Manufacturer:  DWK Life Sciences W266044
 View more versions of this product
Catalog No. 02-911-706
  Check Availability
Add to cart
Description
Description
  • For use with ANTIBind 384- or 96-well plates
  • Packaged in 5 mats/pack
Specifications
Specifications
Sealing Mats
Natural
ANTIBind plates
Silicone with PTFE coating
96
5/Pack