Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldon™ Antibiotic Disks

Determines antibiotic susceptibility of microorganisms.
$8.20
Specifications
Grade | 7 to 14 |
---|---|
Format | Disk |
Diameter (English) Ring | 0.25 in. |
Color | White |
Languages | English |
Description
- Each cartridge contains 50 disks (30mcg) of antibiotics with reclosable cap
- Disks are 6.35mm (0.25 in.) in diameter
- DOT Info: Non-regulated
Specifications
7 to 14 | |
0.25 in. | |
English |
Disk | |
White | |
Biology |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title