Learn More
Raybiotech IncSupplier Diversity Partner C-Series Human Immune Checkpoint Antibody Array 3 Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AAHICM32
Detects 12 Human molecules expressed in APCs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB (TNFRSF9/CD137), B7-H2 (ICOS Ligand), B7-H3 (CD276), B7-H4, CD200, HVEM (TNFRSF14/CD270), CD27 Ligand (TNFSF7), CD80 (B7-1), CD86 (B7-2), OX40 Ligand (TNFSF4), B7-H1 (CD274/PD-L1), PD-L2 (B7-DC)
Catalog No. 50-194-8597