Learn More
Raybiotech Inc C-Series Human Apoptosis Signaling Pathway Array C1.
SDP
Supplier:  Raybiotech Inc AAHAPOSIG18
Encompass_Preferred
Detects 19 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473) (PKB (P-Ser473)), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (Cleaved Asp175), Caspase-7 (Cleaved Asp198), CHK1 (P-Ser296), CHK2 (P-Thr68), eIF-2a (P-Ser52), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183/Tyr185), NFKBP65 (P-Ser536), PARP1 (Cleaved Asp214/Gly215), p27 (P-Thr198), P53 (P-Ser15), SMAD2 (Ser245/250/255), TAK1 (P-Ser412)
Catalog No. 50-194-8482