Learn More
Raybiotech IncSupplier Diversity Partner G-Series Human Neuro Discovery Array 1 Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner GSHNEU11
Semi-Quantitative measurement of 8 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: BDNF, beta-NGF, CNTF, EGF, Eotaxin-1, Eotaxin-2, Eotaxin-3, Fas, GDNF, GM-CSF, IFN-gamma, IL-1 alpha, IL-1 beta, IL-10, IL-18, IL-4, IL-6, IL-8, LIF, MCP-1, MIP-1 beta, MMP-2, MMP-3, RANTES, TARC, TGF beta 1, TIMP-1, TNF alpha, TNF RI, VEGF-A
Catalog No. 50-195-0414