Learn More
BioworldSupplier Diversity Partner Antibody Bio-Stabilizer (10X), 20 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220500051
Antibody Stabilizer is used for long-term storage of diluted antibodies and antibodies conjugates. The Antibody Stabilizer solution provides stability to the proteins/antibodies preserving the native conformation and preventing the loss of functionality due to long storage. Simply dilute the antibody and their conjugate in Antibody Stabilizer solution and store at 2-8 °C.
Catalog No. 50-253-084