Learn More
BioVision IncSupplier Diversity Partner EZLabel™ Antibody Biotin Labeling Kit: Easy to use Kit to label upto 5 mg of Antibody with Biotin for multiple applications such as ELISA, Western blot, Immunohistochemistry, Immunofluorescence and FACS analysis DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K8345
BioVision’s EZLabel™ Antibody-Biotin Labeling Kit provides an easy way to label antibodies with Biotin in a user-friendly spin column format. Due to its high affinity for tetrameric avidin proteins, biotin is ideal for labeling antibodies. Each pair of spin columns provided in the kit can be used to purify up to 1 mg of the labeled target antibody. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. The biotin-labeled antibody can be directly used for multiple downstream applications including ELISA, western blot, Immunohistochemistry, Immunoassays, etc. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9948