Novus Biologicals Antibody Concentration and Clean Up Antibody Purification Kit
Allows quick and easy concentration of antibodies and proteins
Manufacturer: Novus Biologicals 861-0005
 View more versions of this product
Catalog No. 86-100-05
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Antibody Concentration and Clean Up Antibody Purification Kit
    Quantity 1 Column
    The antibody clean up kit method utilizes a simple spin column to easily and quickly remove excess buffer from the antibody thereby providing a more concentrated antibody solution. The clean up kit also allows the experimenter to perform a simple buffer exchange to transfer the antibody into a more favorable conjugation buffer. The kit can also be used to reduce the concentration of many unwanted additives often found in antibody formulations such as azide, glycine or tris.
    Recommended Storage
    Store at 4°C. Do not freeze.
  • Description & Specifications
Related Products
Product Content Feedback