Learn More
BioVision IncSupplier Diversity Partner EZLabel™ Antibody Cy5 Labeling Kit: Easy to use Kit to label upto 5 mg of Antibody with Cy5 for multiple applications such as ELISA, Western blot, Immunohistochemistry, Immunofluorescence and FACS analysis DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K8385
BioVision’s EZLabel™ Antibody-Cy5 Labeling Kit provides an easy way to label antibodies with Cy5 in a user-friendly spin column format. Cy5 is an ideal fluorescent dye for labeling of antibodies with its intense signal in the red region of the spectrum. Each pair of spin columns provided in the kit can be used to purify up to 1 mg of the labeled target protein. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. Cy5-labeled antibody has excitation and emission wavelengths at 650 nm and 670 nm respectively, and can be directly used for multiple downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, FACS, etc. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9952