Learn More
BioworldSupplier Diversity Partner Antibody Dilution and Stabilizer Reagent, 10X, 100 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220500072
Specifications:Provided as 10X stock solution.For use in antibody dilution and stabilization.PBS buffered and sterile filtered.
Catalog No. 50-253-089