Learn More
Aat BioquestSupplier Diversity Partner Buccutite™ Rapid PE Antibody Labeling Kit *Microscale Optimized for Labeling 100 ug Antibody Per Reaction* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1310
R-Phycoerythrin (PE) is an orange fluorescent protein which has an excitation wavelength of 565 nm and an emission wavelength of 575 nm.
Catalog No. 50-195-3032