Learn More
Aat BioquestSupplier Diversity Partner ReadiLink™ Rapid iFluor™ 680 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1240
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. These dyes are optimized to have minimal fluorescence quenching effect on proteins and nucleic acids.
Catalog No. 50-195-3024