Learn More
Aat BioquestSupplier Diversity Partner ReadiLink™ Rapid iFluor™ 594 Antibody Labeling Kit *Microscale Optimized for Labeling 50 µg Antibody Per Reaction* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1230
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. iFluor™ 594 dyes have fluorescence excitation and emission maxima of ~590 nm and ~610 nm respectively.
Catalog No. 50-195-3022