Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Antibody Purification on Protein A Column

Buffers to be used when performing antibody purification using immobilized Protein A

Manufacturer:  Alfa Aesar™ J63014QA

Catalog No. AAJ63014QA


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

This kit contains: 1L of 1M Tris-HCl (pH 8.0) and 500mL of 100mM Glycine (pH 3.0).
Specifications

Specifications

Purification Buffer
1 Kit
Liquid
Buffers to be used when performing antibody purification using immobilized Protein A.
Liquid
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit