Learn More
Bel-Art™ SP Scienceware™ Antibody Saver Tray Sets; Disposable/Reusable
Sized to reduce antibody usage in Western blots. The Bel-Art - SP Scienceware Antibody Saver Tray Sets include multiple trays and lanes in two sizes, which can be mixed and matched for maximum efficiency when performing Western blots.
Manufacturer:  Bel-Art™ F451000020
 View more versions of this product
Catalog No. 03-421-176
  Check Availability
Add to cart
Specifications
Description
  • Reduce antibody volume by using only the tray/lane(s) needed 
  • Sets contain full and half blot trays as well as single lane, 2 lane, or 3 lane (Midi only) 
  • Included lid protects samples and allows stacking
  • Dispose or reuse based on your needs
  • Transparent PVC
Description & Specifications
Specifications
7.25 in. x 5.875 in. (18.5cm x 15cm)
3 sets - Set includes full blot (1), half blot (2), 3 lane (3), 2 lane (4), single lane (6) and lid
Transparent PVC
For midi blots