Learn More
Contec™ Anticon™ MicroQuilt Wipes
Contec™ Anticon™ MicroQuilt Wipes combine the cleanliness and sorbent capacity of Quiltec™ with the microfiber particle pick-up ability of Anticon™ Gold Sorb.
Manufacturer:  Contec™ 495758736
 View more versions of this product
Catalog No. 19-168-577
Add to cart
Description
Description
  • Laminated 2-ply wipes
  • Stronger and more absorbent wipe
  • Quilted pockets between the fabric layers, increase the sorbent capacity
  • Readily releases fluid to a surface when required
  • Unique microfiber polyester yarn
  • Provides superior residue and particle removal even when dry
  • High sorptive capacity and fast absorption rate for superior spill control
  • Good solvent compatibility
  • Ideal for use with a wide range of disinfectants and solutions
  • Sealed edges reduce particle and fiber generation, reducing possibility of contamination
Specifications
Specifications
9 x 9 in. (22.86 x 22.86 cm)
9 in.
Polyester, microfiber
Sealed
25 wipes/bag, 40 bags/case
9 in.
Sterile
Cleanroom applications requiring high sorbent capacity and superior contaminant removal
22.86 cm
Wipe
Flat stacked
2-ply
22.86 cm