Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antifoam AF, Emulsion, Food Grade, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Cubitainer
Drum
Poly Bottle
Description
Spectrum™ Chemical Antifoam AF, Emulsion, Food Grade is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
CAS Max % | 100% |
Quantity | 4 L |
Grade | Food Grade |
Packaging | Poly Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction