Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Antimony, AAS standard solution, Specpure™, Sb 1000µg/mL

Manufacturer:  Alfa Aesar™ 33558AP

Quantity 500mL

 View more versions of this product

Catalog No. AA33558AP

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

MFCD00134030
UN1789
Antimony AAS standard solution, Sb 1000μg/mL
Specpure™

Specifications

Liquid
MFCD00134030
500mL
Specpure™
Sb in 20% HCl
UN1789
Antimony AAS standard solution, Sb 1000μg/mL

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit