Alfa Aesar™ Antimony(III) bromide, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 1168618

Quantity 50g

 View more versions of this product

Catalog No. AA1168618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-61-9
    Formula Weight 361.48
    MDL Number MFCD00016317
    UN Number UN3260
    Merck Index 14,706
    Chemical Name or Material Antimony(III) bromide

    Specifications

    Formula Weight 361.48
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Characteristic
    MDL Number MFCD00016317
    UN Number UN3260
    Merck Index 14,706
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Physical Form Crystalline
    Refractive Index 1.74
    Sensitivity Moisture Sensitive
    CAS 7789-61-9
    Boiling Point 280°C
    Melting Point 96.6°C
    Quantity 50g
    Health Hazard 1 Warning
    Density 4.148g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Antimony(III) bromide
    Assay 99.995%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit