Alfa Aesar™ Antimony(III) iodide, ultra dry, 99.998% (metals basis)

Manufacturer: Alfa Aesar™ 3567606

Quantity 5g

 View more versions of this product

Catalog No. AA3567606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-44-5
    Formula Weight 502.46
    MDL Number MFCD00016322
    UN Number UN3260
    Merck Index 14,710
    Chemical Name or Material Antimony(III) iodide, Ultra dry

    Specifications

    Formula Weight 502.46
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,710
    Physical Form Beads
    CAS 7790-44-5
    UN Number UN3260
    Quantity 5g
    Solubility Information Decomposed by water to SbOI
    Odor Acrid
    Sensitivity Moisture Sensitive
    Boiling Point 420°C
    Melting Point 168°C
    Health Hazard 1 Warning
    Storage Note 1
    • Ampouled under argon
    TSCA TSCA
    Mesh Size −10 Mesh
    MDL Number MFCD00016322
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Antimony(III) iodide, Ultra dry
    Assay 99.998%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit