Alfa Aesar™ Antimony(III) methoxide, 95%

Manufacturer: Alfa Aesar™ 3349504

Quantity 2g

 View more versions of this product

Catalog No. AA3349504

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 29671-18-9
    Formula Weight 214.8
    MDL Number MFCD00014882
    UN Number UN3181
    Chemical Name or Material Antimony(III) methoxide

    Specifications

    Formula Weight 214.8
    Physical Form Crystalline
    MDL Number MFCD00014882
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Sensitivity Air & Moisture Sensitive
    UN Number UN3181
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Melting Point 124°C
    Quantity 2g
    Flash Point >110° (230°F)
    CAS 29671-18-9
    Health Hazard 3 GHS P Statement
    P273
    Avoid release to the environment.
    Chemical Name or Material Antimony(III) methoxide
    Assay 95%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273
    Avoid release to the environment.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit