Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Antimony(III) n-butoxide, 99+%

Preparation of coatings, as a catalyst

Manufacturer:  Alfa Aesar™ 4329509

Quantity 10g

 View more versions of this product

Catalog No. AA4329509


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2155-74-0
341.1
UN3082
Sb(OC4H9)3
MFCD00015238
Antimony (III) n-butoxide

Specifications

1.264g/mL
133° to 135°C (0.4mmHg)
>110°C (230°F)
Liquid
341.1
UN3082
Antimony (III) n-butoxide
2155-74-0
10g
Moisture Sensitive
Sb(OC4H9)3
MFCD00015238
Soluble in anhydrous butanol
≥99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273
Avoid release to the environment.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit