Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antimony(III) oxide, 99+%, ACROS Organics™
Click to view available options
Quantity:
100g
1kg
5kg
Specifications
Assay Percent Range | 99% min. (Sb2O3) |
Linear Formula | Sb2O3 |
Merck Index | 15,699 |
Solubility Information | Solubility in water: 2.7mg/l (20°C). Other solubilities: slightly soluble in hot water,soluble in conc. hcl,acetic acid,sulfuric acids,,solutions of alkali hydroxides |
Formula Weight | 291.42 |
Percent Purity | 99+% |
Packaging | Plastic Bottle |
Boiling Point | 1550°C |
Melting Point | 656°C |
Quantity | 1kg |
Show More |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 215-175-