Alfa Aesar™ Antimony(III) sulfide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 4178230

Quantity 250g

 View more versions of this product

Catalog No. AA4178230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1345-04-6
    Formula Weight 339.68
    MDL Number MFCD00011217
    UN Number UN1549
    Merck Index 14,713
    Chemical Name or Material Antimony(III) sulfide

    Specifications

    Formula Weight 339.68
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size 25mm & down
    Physical Form Pieces
    UN Number UN1549
    Merck Index 14,713
    Melting Point 550°C
    Health Hazard 1 Warning
    TSCA TSCA
    MDL Number MFCD00011217
    CAS 1345-04-6
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
    Quantity 250g
    Chemical Name or Material Antimony(III) sulfide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit