Alfa Aesar™ Antimony(III) telluride, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4592218

Quantity 50g

 View more versions of this product

Catalog No. AA4592218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1327-50-0
    Molecular Formula Sb2Te3
    Formula Weight 626.3
    MDL Number MFCD00016320
    UN Number UN1549
    Chemical Name or Material Antimony (III) telluride

    Specifications

    Formula Weight 626.3
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1549
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    MDL Number MFCD00016320
    CAS 1327-50-0
    Color Gray
    Density 6.5g/mL
    Mesh Size −100 Mesh
    Odor Odorless
    Physical Form Powder
    Molecular Formula Sb2Te3
    Melting Point 629°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
    Chemical Name or Material Antimony (III) telluride
    Assay 99.999%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit